Product Details

SNP ID
rs192376885
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:76471765 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACAGCACGAGGGCGGCGAAGAGGCC[A/G]GCAAAGGCCAGCAGTGACACCAGGA
Phenotype
MIM: 600950 MIM: 614404
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
AANAT PubMed Links

Gene Details

Gene
AANAT
Gene Name
aralkylamine N-acetyltransferase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001088.2 3094 Intron NP_001079.1
NM_001166579.1 3094 Intron NP_001160051.1
XM_017024259.1 3094 Intron XP_016879748.1
Gene
RHBDF2
Gene Name
rhomboid 5 homolog 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001005498.3 3094 Silent Mutation GCC,GCT A784A NP_001005498.2
NM_024599.5 3094 Silent Mutation GCC,GCT A813A NP_078875.4
XM_005257669.3 3094 Silent Mutation GCC,GCT A813A XP_005257726.1
XM_005257670.1 3094 Silent Mutation GCC,GCT A784A XP_005257727.1
XM_011525249.2 3094 Silent Mutation GCC,GCT A784A XP_011523551.1
XM_011525250.2 3094 Silent Mutation GCC,GCT A784A XP_011523552.1
XM_011525251.2 3094 Silent Mutation GCC,GCT A784A XP_011523553.1
XM_017025079.1 3094 Silent Mutation GCC,GCT A784A XP_016880568.1

View Full Product Details