Product Details

SNP ID
rs7913883
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:68724609 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTAGCCAGTTGTTTGAATTTAAAGT[C/T]GAAAAACTACATTCAAAAAAAAAAA
Phenotype
MIM: 612569
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CCAR1 PubMed Links

Gene Details

Gene
CCAR1
Gene Name
cell division cycle and apoptosis regulator 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001282959.1 Intron NP_001269888.1
NM_001282960.1 Intron NP_001269889.1
NM_018237.3 Intron NP_060707.2

View Full Product Details