Product Details

SNP ID
rs200279130
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:98395276 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAGTGGCTTTTAAAGGCTCAGACTC[A/G]AACCACTGATCCAGCACCTGGACAG
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
MIR1287 PubMed Links

Gene Details

Gene
MIR1287
Gene Name
microRNA 1287
There are no transcripts associated with this gene.

Gene
PYROXD2
Gene Name
pyridine nucleotide-disulphide oxidoreductase domain 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_032709.2 752 Silent Mutation TTC,TTT F235F NP_116098.2
XM_011540293.2 752 Silent Mutation TTC,TTT F269F XP_011538595.1
XM_011540301.2 752 Intron XP_011538603.1
XM_017016835.1 752 Silent Mutation TTC,TTT F235F XP_016872324.1
XM_017016836.1 752 Silent Mutation TTC,TTT F235F XP_016872325.1
XM_017016837.1 752 Silent Mutation TTC,TTT F235F XP_016872326.1
XM_017016838.1 752 Silent Mutation TTC,TTT F114F XP_016872327.1
XM_017016839.1 752 Silent Mutation TTC,TTT F114F XP_016872328.1
XM_017016840.1 752 Silent Mutation TTC,TTT F114F XP_016872329.1
XM_017016841.1 752 Silent Mutation TTC,TTT F114F XP_016872330.1

View Full Product Details