Product Details
- SNP ID
-
rs200107165
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.10:127738722 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TGCAGGCCTCGCCCTCTCCATCCGC[A/T]CCCGAGGCCGCCACCTGCTTCTCCG
- Phenotype
-
- Polymorphism
- A/T, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
FOXI2
PubMed Links
Gene Details
- Gene
- FOXI2
- Gene Name
- forkhead box I2
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_207426.2 |
714 |
Silent Mutation |
GCA,GCT |
A238A |
NP_997309.2 |
- Gene
- LOC387720
- Gene Name
- collagen alpha-1(X) chain
There are no transcripts associated with this gene.
View Full Product Details