Product Details

SNP ID
rs201985510
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:89206498 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATCACCGCGCTGGGACAGATGCAGT[C/T]CGCAGCGTTCCCAGTATTTTGTCCC
Phenotype
MIM: 604551 MIM: 613497
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CH25H PubMed Links

Gene Details

Gene
CH25H
Gene Name
cholesterol 25-hydroxylase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003956.3 817 Silent Mutation CGA,CGG R265R NP_003947.1
Gene
LIPA
Gene Name
lipase A, lysosomal acid type
There are no transcripts associated with this gene.

View Full Product Details