Product Details

SNP ID
rs201417018
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:50086199 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGCCAGGCAGCCGAATGACACCTCA[C/G]GGACCTTCCATGGGACCCCCTGGCT
Phenotype
MIM: 602866 MIM: 601735
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
ASIC1 PubMed Links

Gene Details

Gene
ASIC1
Gene Name
acid sensing ion channel subunit 1
There are no transcripts associated with this gene.

Gene
SMARCD1
Gene Name
SWI/SNF related, matrix associated, actin dependent regulator of chromatin, subfamily d, member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003076.4 386 Missense Mutation CAC,CAG H72Q NP_003067.3
NM_139071.2 386 Missense Mutation CAC,CAG H72Q NP_620710.2
XM_005269107.3 386 Missense Mutation CAC,CAG H72Q XP_005269164.2
XM_011538695.1 386 Missense Mutation CAC,CAG H72Q XP_011536997.1

View Full Product Details