Product Details

SNP ID
rs200843733
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.13:39039617 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGTTCACAGAGGATGGATATTTCCT[A/G]CGAGCCTGGAATTATACAGTTGACA
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
NHLRC3 PubMed Links

Gene Details

Gene
NHLRC3
Gene Name
NHL repeat containing 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001012754.3 620 Silent Mutation CTA,CTG L97L NP_001012772.1
NM_001017370.2 620 Silent Mutation CTA,CTG L97L NP_001017370.1
Gene
PROSER1
Gene Name
proline and serine rich 1
There are no transcripts associated with this gene.

View Full Product Details