Product Details
- SNP ID
-
rs202011663
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.13:41223350 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AATTCGCTCTGACTGGGCTCTTGTT[C/G]CCACCTGTGCCATAACAAAAAGCAA
- Phenotype
-
MIM: 604601
- Polymorphism
- C/G, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
LOC101929140
PubMed Links
Gene Details
- Gene
- LOC101929140
- Gene Name
- uncharacterized LOC101929140
There are no transcripts associated with this gene.
- Gene
- MTRF1
- Gene Name
- mitochondrial translational release factor 1
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_004294.2 |
1670 |
Missense Mutation |
GCA,GGA |
A377G |
NP_004285.2 |
XM_005266599.2 |
1670 |
Missense Mutation |
GCA,GGA |
A377G |
XP_005266656.1 |
XM_006719897.2 |
1670 |
Missense Mutation |
GCA,GGA |
A377G |
XP_006719960.1 |
XM_006719898.2 |
1670 |
Missense Mutation |
GCA,GGA |
A377G |
XP_006719961.1 |
XM_006719900.2 |
1670 |
Missense Mutation |
GCA,GGA |
A377G |
XP_006719963.1 |
XM_011535315.2 |
1670 |
Missense Mutation |
GCA,GGA |
A429G |
XP_011533617.1 |
XM_011535317.2 |
1670 |
Missense Mutation |
GCA,GGA |
A390G |
XP_011533619.1 |
XM_011535318.2 |
1670 |
Missense Mutation |
GCA,GGA |
A377G |
XP_011533620.1 |
XM_011535320.2 |
1670 |
Missense Mutation |
GCA,GGA |
A377G |
XP_011533622.1 |
XM_011535321.2 |
1670 |
Missense Mutation |
GCA,GGA |
A377G |
XP_011533623.1 |
XM_011535323.2 |
1670 |
Missense Mutation |
GCA,GGA |
A377G |
XP_011533625.1 |
XM_011535325.2 |
1670 |
Missense Mutation |
GCA,GGA |
A377G |
XP_011533627.1 |
XM_011535326.2 |
1670 |
Intron |
|
|
XP_011533628.1 |
XM_011535327.2 |
1670 |
Intron |
|
|
XP_011533629.1 |
XM_017020856.1 |
1670 |
Missense Mutation |
GCA,GGA |
A429G |
XP_016876345.1 |
XM_017020857.1 |
1670 |
Missense Mutation |
GCA,GGA |
A429G |
XP_016876346.1 |
XM_017020858.1 |
1670 |
Missense Mutation |
GCA,GGA |
A429G |
XP_016876347.1 |
XM_017020859.1 |
1670 |
Missense Mutation |
GCA,GGA |
A429G |
XP_016876348.1 |
XM_017020860.1 |
1670 |
Missense Mutation |
GCA,GGA |
A390G |
XP_016876349.1 |
XM_017020861.1 |
1670 |
Missense Mutation |
GCA,GGA |
A390G |
XP_016876350.1 |
XM_017020862.1 |
1670 |
Missense Mutation |
GCA,GGA |
A377G |
XP_016876351.1 |
XM_017020863.1 |
1670 |
Missense Mutation |
GCA,GGA |
A377G |
XP_016876352.1 |
XM_017020864.1 |
1670 |
Missense Mutation |
GCA,GGA |
A377G |
XP_016876353.1 |
XM_017020865.1 |
1670 |
Missense Mutation |
GCA,GGA |
A377G |
XP_016876354.1 |
XM_017020866.1 |
1670 |
Missense Mutation |
GCA,GGA |
A377G |
XP_016876355.1 |
XM_017020867.1 |
1670 |
Missense Mutation |
GCA,GGA |
A377G |
XP_016876356.1 |
XM_017020868.1 |
1670 |
Missense Mutation |
GCA,GGA |
A377G |
XP_016876357.1 |
XM_017020869.1 |
1670 |
Missense Mutation |
GCA,GGA |
A377G |
XP_016876358.1 |
XM_017020870.1 |
1670 |
Missense Mutation |
GCA,GGA |
A377G |
XP_016876359.1 |
XM_017020871.1 |
1670 |
Missense Mutation |
GCA,GGA |
A377G |
XP_016876360.1 |
XM_017020872.1 |
1670 |
Intron |
|
|
XP_016876361.1 |
XM_017020873.1 |
1670 |
Missense Mutation |
GCA,GGA |
A377G |
XP_016876362.1 |
XM_017020874.1 |
1670 |
Missense Mutation |
GCA,GGA |
A377G |
XP_016876363.1 |
XM_017020875.1 |
1670 |
Missense Mutation |
GCA,GGA |
A377G |
XP_016876364.1 |
XM_017020876.1 |
1670 |
Missense Mutation |
GCA,GGA |
A213G |
XP_016876365.1 |
XM_017020877.1 |
1670 |
Missense Mutation |
GCA,GGA |
A213G |
XP_016876366.1 |
XM_017020878.1 |
1670 |
Intron |
|
|
XP_016876367.1 |
XM_017020879.1 |
1670 |
Intron |
|
|
XP_016876368.1 |
View Full Product Details