Product Details

SNP ID
rs200859196
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:60509558 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGCCCTCAACAAGAATGAGTCGGTG[C/G]TACGCGCACGAGCCATCGTGGCCTT
Phenotype
MIM: 606326
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
C14orf39 PubMed Links

Gene Details

Gene
C14orf39
Gene Name
chromosome 14 open reading frame 39
There are no transcripts associated with this gene.

Gene
SIX6
Gene Name
SIX homeobox 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_007374.2 339 Missense Mutation CTA,GTA L54V NP_031400.2

View Full Product Details