Product Details

SNP ID
rs201343049
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.14:20242994 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGATGCAACCCACTACTACACACCC[C/G]CATGTACTTTCTGCTGGGAAATTTT
Phenotype
Polymorphism
C/G, Transversion Substitution
Allele Nomenclature
Literature Links
OR11H4 PubMed Links

Gene Details

Gene
OR11H4
Gene Name
olfactory receptor family 11 subfamily H member 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001004479.1 203 Missense Mutation CCC,CGC P68R NP_001004479.1

View Full Product Details