Product Details

SNP ID
rs200419591
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:21665780 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTTCGAAAACACTCAGCTGAAGGGC[A/G]CCGGAAAGCCCTGTCTACCTGCTCG
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
OR4E1 PubMed Links

Gene Details

Gene
OR4E1
Gene Name
olfactory receptor family 4 subfamily E member 1 (gene/pseudogene)
There are no transcripts associated with this gene.

Gene
OR4E2
Gene Name
olfactory receptor family 4 subfamily E member 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001001912.2 698 Missense Mutation CAC,CGC H233R NP_001001912.2

View Full Product Details