Product Details

SNP ID
rs200319330
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:75112475 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGCAGCAGCTGCCGCTTGCCCATTC[C/T]GATGCCATGCTCCCCCGTGCACGTT
Phenotype
MIM: 607490 MIM: 612060
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LDHD PubMed Links

Gene Details

Gene
LDHD
Gene Name
lactate dehydrogenase D
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_153486.3 1388 Missense Mutation AGA,GGA R469G NP_705690.2
NM_194436.2 1388 Missense Mutation AGA,GGA R446G NP_919417.1
Gene
ZNRF1
Gene Name
zinc and ring finger 1, E3 ubiquitin protein ligase
There are no transcripts associated with this gene.

View Full Product Details