Product Details

SNP ID
rs202233631
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:70251264 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGATTCAGCACAGGCATGAGCGCCA[C/T]GGTGAGGCCGGCGGCGGCGCGCTCT
Phenotype
MIM: 601065 MIM: 606490
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
AARS PubMed Links

Gene Details

Gene
AARS
Gene Name
alanyl-tRNA synthetase
There are no transcripts associated with this gene.

Gene
EXOSC6
Gene Name
exosome component 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_058219.2 667 Missense Mutation ATG,GTG M213V NP_478126.1
Gene
LOC107984138
Gene Name
serine/threonine-protein kinase SMG1-like
There are no transcripts associated with this gene.

View Full Product Details