Product Details

SNP ID
rs199834493
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:40999844 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGGACACTGCAGCTGCGAGAGGCAC[A/G]GCAATCCATGGCAATGGGCGTCTGG
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
KRTAP3-1 PubMed Links

Gene Details

Gene
KRTAP3-1
Gene Name
keratin associated protein 3-1
There are no transcripts associated with this gene.

Gene
KRTAP3-2
Gene Name
keratin associated protein 3-2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_031959.2 43 Missense Mutation CGT,TGT R4C NP_114165.1
Gene
KRTAP3-3
Gene Name
keratin associated protein 3-3
There are no transcripts associated with this gene.

View Full Product Details