Product Details

SNP ID
rs199962683
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:75519031 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCTGAAGAGGTTGGGTTATGTGGTT[C/T]GACGATTCCAACCAAGGTAAATCCC
Phenotype
MIM: 612185 MIM: 608755
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CASKIN2 PubMed Links

Gene Details

Gene
CASKIN2
Gene Name
CASK interacting protein 2
There are no transcripts associated with this gene.

Gene
LLGL2
Gene Name
LLGL2, scribble cell polarity complex component
There are no transcripts associated with this gene.

Gene
TSEN54
Gene Name
tRNA splicing endonuclease subunit 54
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_207346.2 537 Nonsense Mutation CGA,TGA R169* NP_997229.2
XM_005257229.3 537 Nonsense Mutation CGA,TGA R169* XP_005257286.1

View Full Product Details