Product Details

SNP ID
rs199760099
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:12319471 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCCACCTTGCACTACCATGTGTCTT[C/T]GAAGGTTTCTACGAGAACTGAAGGT
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ZNF563 PubMed Links

Gene Details

Gene
ZNF563
Gene Name
zinc finger protein 563
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_145276.2 792 Missense Mutation CAA,CGA Q185R NP_660319.1
XM_005259750.4 792 Intron XP_005259807.1
XM_005259751.3 792 Intron XP_005259808.1
XM_006722650.3 792 Intron XP_006722713.1
XM_006722651.3 792 Intron XP_006722714.1
XM_011527698.1 792 Intron XP_011526000.1
XM_011527699.2 792 Missense Mutation CAA,CGA Q148R XP_011526001.1
XM_011527700.2 792 Intron XP_011526002.1
XM_017026332.1 792 Intron XP_016881821.1

View Full Product Details