Product Details

SNP ID
rs200434887
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:18589507 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCACCGCCAGTACCTGCGCAGCGGC[C/G]CTGACTACGACTTCGCGCGCTACCG
Phenotype
MIM: 604237
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
C19orf60 PubMed Links

Gene Details

Gene
C19orf60
Gene Name
chromosome 19 open reading frame 60
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001100418.1 315 Missense Mutation CCT,GCT P93A NP_001093888.1
NM_001100419.1 315 Missense Mutation CCT,GCT P71A NP_001093889.1
XM_005259977.1 315 Missense Mutation CCT,GCT P105A XP_005260034.1
Gene
CRLF1
Gene Name
cytokine receptor like factor 1
There are no transcripts associated with this gene.

View Full Product Details