Product Details

SNP ID
rs199878965
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:12319588 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTGATGGTAACTGAAGGCTTTCCCA[C/T]GTTGTTTATGTGTATGTGGCTTCTC
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ZNF563 PubMed Links

Gene Details

Gene
ZNF563
Gene Name
zinc finger protein 563
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_145276.2 675 Missense Mutation CAT,CGT H146R NP_660319.1
XM_005259750.4 675 Intron XP_005259807.1
XM_005259751.3 675 Intron XP_005259808.1
XM_006722650.3 675 Intron XP_006722713.1
XM_006722651.3 675 Intron XP_006722714.1
XM_011527698.1 675 Intron XP_011526000.1
XM_011527699.2 675 Missense Mutation CAT,CGT H109R XP_011526001.1
XM_011527700.2 675 Intron XP_011526002.1
XM_017026332.1 675 Intron XP_016881821.1

View Full Product Details