Product Details

SNP ID
rs199703729
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:56516289 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGTCACTTCAGGACTTAGTGACATT[C/T]AAGGATGTGGCAATAGATTTTTCCC
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ZNF471 PubMed Links

Gene Details

Gene
ZNF471
Gene Name
zinc finger protein 471
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001321768.1 181 Intron NP_001308697.1
NM_020813.3 181 Silent Mutation TTC,TTT F16F NP_065864.2
XM_011527148.1 181 Silent Mutation TTC,TTT F16F XP_011525450.1
XM_017027032.1 181 Intron XP_016882521.1

View Full Product Details