Product Details

SNP ID
rs202169219
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:12318655 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTGACATACACTGGGATAAACAAAG[C/G]CTTTCCCACATACCTTGCATTTATT
Phenotype
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
ZNF563 PubMed Links

Gene Details

Gene
ZNF563
Gene Name
zinc finger protein 563
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_145276.2 1608 Missense Mutation GCC,GGC A457G NP_660319.1
XM_005259750.4 1608 Intron XP_005259807.1
XM_005259751.3 1608 Intron XP_005259808.1
XM_006722650.3 1608 Intron XP_006722713.1
XM_006722651.3 1608 Intron XP_006722714.1
XM_011527698.1 1608 Intron XP_011526000.1
XM_011527699.2 1608 Missense Mutation GCC,GGC A420G XP_011526001.1
XM_011527700.2 1608 Intron XP_011526002.1
XM_017026332.1 1608 Intron XP_016881821.1

View Full Product Details