Product Details

SNP ID
rs200573717
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:14093637 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGGAGGGGAGGCCCACCTTCCTCTG[A/G]TAGATGGCAATCCAGTCAGTTGTGG
Phenotype
MIM: 601639
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
C19orf67 PubMed Links

Gene Details

Gene
C19orf67
Gene Name
chromosome 19 open reading frame 67
There are no transcripts associated with this gene.

Gene
PRKACA
Gene Name
protein kinase cAMP-activated catalytic subunit alpha
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001304349.1 1555 Silent Mutation TAC,TAT Y383Y NP_001291278.1
NM_002730.3 1555 Silent Mutation TAC,TAT Y307Y NP_002721.1
NM_207518.2 1555 Intron NP_997401.1
XM_017026948.1 1555 Intron XP_016882437.1
Gene
SAMD1
Gene Name
sterile alpha motif domain containing 1
There are no transcripts associated with this gene.

View Full Product Details