Product Details

SNP ID
rs201135202
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:18596764 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTTTCAGGCCGGCCAGGCGGCAGGA[C/G]GTCTGGTTGCTCACATCGTCCACCA
Phenotype
MIM: 604237
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
C19orf60 PubMed Links

Gene Details

Gene
C19orf60
Gene Name
chromosome 19 open reading frame 60
There are no transcripts associated with this gene.

Gene
CRLF1
Gene Name
cytokine receptor like factor 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004750.4 1150 Silent Mutation ACC,ACG T294T NP_004741.1
XM_011528422.2 1150 Silent Mutation ACC,ACG T272T XP_011526724.1
XM_011528423.2 1150 Silent Mutation ACC,ACG T294T XP_011526725.1
XM_011528424.2 1150 Silent Mutation ACC,ACG T272T XP_011526726.1

View Full Product Details