Product Details

SNP ID
rs200591095
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.21:40013056 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGTTATACCAGGGTGTAAGATTTT[C/G]CGTAAGGATTGTTTCCTTTCAAATG
Phenotype
MIM: 602523
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
DSCAM PubMed Links

Gene Details

Gene
DSCAM
Gene Name
DS cell adhesion molecule
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001271534.1 6469 Missense Mutation GCA,GGA A1988G NP_001258463.1
NM_001389.3 6469 Missense Mutation GCA,GGA A2006G NP_001380.2
XM_017028281.1 6469 Missense Mutation GCA,GGA A1770G XP_016883770.1

View Full Product Details