Product Details

SNP ID
rs200198659
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.21:40013087 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGATTGTTTCCTTTCAAATGGCCCC[A/G]GGAGTCGAGCAGTGATTCTTGGGAG
Phenotype
MIM: 602523
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
DSCAM PubMed Links

Gene Details

Gene
DSCAM
Gene Name
DS cell adhesion molecule
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001271534.1 6438 Missense Mutation CGG,TGG R1978W NP_001258463.1
NM_001389.3 6438 Missense Mutation CGG,TGG R1996W NP_001380.2
XM_017028281.1 6438 Missense Mutation CGG,TGG R1760W XP_016883770.1

View Full Product Details