Product Details

SNP ID
rs200705717
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:85694447 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCTACCTGGGCCCTCCTGCTCCTTG[C/T]AGCCATGCTCCTGGGCAACCCAGGT
Phenotype
MIM: 188855
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
GNLY PubMed Links

Gene Details

Gene
GNLY
Gene Name
granulysin
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001302758.1 90 Missense Mutation GCA,GTA A10V NP_001289687.1
NM_006433.4 90 Missense Mutation GCA,GTA A10V NP_006424.2
NM_012483.3 90 UTR 5 NP_036615.2
XM_005264084.2 90 Missense Mutation GCA,GTA A10V XP_005264141.1
XM_005264085.2 90 Missense Mutation GCA,GTA A10V XP_005264142.1
XM_005264087.2 90 UTR 5 XP_005264144.1

View Full Product Details