Product Details
- SNP ID
-
rs201184651
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.3:46358344 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CCTGAACACCTTCCAGGAATTCTTC[A/G]GCCTGAGTAACTGTGAAAGCACCAG
- Phenotype
-
MIM: 601267
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
CCR2
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs1799865] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- CCR2
- Gene Name
- C-C motif chemokine receptor 2
- Gene
- LOC102724297
- Gene Name
- uncharacterized LOC102724297
There are no transcripts associated with this gene.
View Full Product Details