Product Details

SNP ID
rs200888528
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:99513208 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCCCAGAAGAGTCTGCCACATCAAA[A/G]GTAAGGTGACTTAAGGTCCTAGTAT
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
C4orf17 PubMed Links

Gene Details

Gene
C4orf17
Gene Name
chromosome 4 open reading frame 17
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_032149.2 489 Missense Mutation AGC,GGC S43G NP_115525.2
XM_011532315.1 489 Missense Mutation AGC,GGC S43G XP_011530617.1

View Full Product Details