Product Details

SNP ID
rs201706642
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:185405781 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAAACAGTGTCTGAAACAGCTTACC[C/G]ATCATAACTGGAGTTCCTTCACTTT
Phenotype
MIM: 611482
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
ANKRD37 PubMed Links

Gene Details

Gene
ANKRD37
Gene Name
ankyrin repeat domain 37
There are no transcripts associated with this gene.

Gene
C4orf47
Gene Name
chromosome 4 open reading frame 47
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001114357.1 1314 Intron NP_001107829.1
XM_005263019.3 1314 Intron XP_005263076.1
XM_011531989.2 1314 Intron XP_011530291.1
XM_017008236.1 1314 Intron XP_016863725.1
XM_017008237.1 1314 Intron XP_016863726.1
XM_017008238.1 1314 Intron XP_016863727.1
XM_017008239.1 1314 Intron XP_016863728.1
XM_017008240.1 1314 Intron XP_016863729.1
XM_017008241.1 1314 Intron XP_016863730.1
XM_017008242.1 1314 Intron XP_016863731.1
Gene
LRP2BP
Gene Name
LRP2 binding protein
There are no transcripts associated with this gene.

Gene
UFSP2
Gene Name
UFM1 specific peptidase 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_018359.3 1314 Silent Mutation ATC,ATG I399M NP_060829.2

View Full Product Details