Product Details
- SNP ID
-
rs201368502
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.4:99513146 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GGCAAAGGCAGCCATATTATGGCTA[A/G]AAATGTAAGCTGCTTTCTAGTCAGG
- Phenotype
-
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
C4orf17
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs554111966] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- C4orf17
- Gene Name
- chromosome 4 open reading frame 17
View Full Product Details