Product Details

SNP ID
rs201755131
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:127707082 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGGCAGGCAAGGAGCTTGAAGTCAA[A/G]AAGAAAGGGGGCAAGAAGGAGCCGG
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CFAP157 PubMed Links

Gene Details

Gene
CFAP157
Gene Name
cilia and flagella associated protein 157
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001012502.2 94 Silent Mutation AAA,AAG K17K NP_001012520.2
XM_006717064.3 94 Silent Mutation AAA,AAG K112K XP_006717127.2
XM_011518559.2 94 Silent Mutation AAA,AAG K112K XP_011516861.2
XM_017014629.1 94 Silent Mutation AAA,AAG K112K XP_016870118.1
XM_017014630.1 94 UTR 5 XP_016870119.1
Gene
PTRH1
Gene Name
peptidyl-tRNA hydrolase 1 homolog
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001002913.1 94 Intron NP_001002913.1
XM_006716955.3 94 Intron XP_006717018.1
XM_017014276.1 94 Intron XP_016869765.1
XM_017014277.1 94 Intron XP_016869766.1
XM_017014278.1 94 Intron XP_016869767.1
Gene
TTC16
Gene Name
tetratricopeptide repeat domain 16
There are no transcripts associated with this gene.

View Full Product Details