Product Details
- SNP ID
-
rs2269488
- Assay Type
- Validated
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.4:2469165 on Build GRCh38
- Set Membership
-
Validated
- Context Sequence [VIC/FAM]
- TTTCTCGAGGAGCTCTCCTGGGCGG[C/G]TGAAGAAGGAGCTTCTTCTCCGGAG
- Phenotype
-
MIM: 602850
- Polymorphism
- C/G, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
CFAP99
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs71606399] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- CFAP99
- Gene Name
- cilia and flagella associated protein 99
There are no transcripts associated with this gene.
- Gene
- RNF4
- Gene Name
- ring finger protein 4
View Full Product Details