Product Details

SNP ID
rs8029246
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.15:45528960 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
AATACAATAAAAAGTCACATGAAAC[A/G]TCATTTTCCTCTGAAGATTGAAATG
Phenotype
MIM: 611314 MIM: 602095
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
HMGN2P46 PubMed Links

Gene Details

Gene
HMGN2P46
Gene Name
high mobility group nucleosomal binding domain 2 pseudogene 46
There are no transcripts associated with this gene.

Gene
SLC30A4
Gene Name
solute carrier family 30 member 4
There are no transcripts associated with this gene.

View Full Product Details