Product Details
- SNP ID
-
rs1129609
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.8:120535940 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GAAAACAGGGTCCAGAAAGGCCCTG[A/C]CCATTAATTTTAAAACTTTCTGACC
- Phenotype
-
MIM: 600026
- Polymorphism
- A/C, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
SNTB1
PubMed Links
Gene Details
- Gene
- SNTB1
- Gene Name
- syntrophin beta 1
View Full Product Details