Product Details
- SNP ID
-
hCV11616140
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.X:38154576 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GATGTGTCGAAGATCAAGGCCACAC[A/T]GTGCTTGCTGGGAAAAAGAAAACCC
- Phenotype
-
MIM: 300187
- Polymorphism
- A/T, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
SRPX
PubMed Links
Gene Details
- Gene
- SRPX
- Gene Name
- sushi repeat containing protein, X-linked
There are no transcripts associated with this gene.
View Full Product Details