Product Details

SNP ID
hCV12123380
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:119140923 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACGGTGGACTCTGTGTTCTTTCTTC[A/G]AATCCTCAACTTGAGACAGGTTTAT
Phenotype
MIM: 615564
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
FAM45A PubMed Links

Gene Details

Gene
FAM45A
Gene Name
family with sequence similarity 45 member A
There are no transcripts associated with this gene.

Gene
SFXN4
Gene Name
sideroflexin 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_213649.1 1378 UTR 3 NP_998814.1

View Full Product Details