Product Details

SNP ID
rs2070818
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:154380798 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GTCTTCTTCTGAAGAGGAGTGCAAG[C/G]ATAGGTGCGTAGTGGGGGAGCCCAG
Phenotype
MIM: 300384 MIM: 300017
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
EMD PubMed Links

Gene Details

Gene
EMD
Gene Name
emerin
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000117.2 693 Missense Mutation CAT,GAT H149D NP_000108.1
Gene
FLNA
Gene Name
filamin A
There are no transcripts associated with this gene.

Gene
LOC107987332
Gene Name
uncharacterized LOC107987332
There are no transcripts associated with this gene.

View Full Product Details