Product Details
- SNP ID
-
rs2289801
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.17:76471082 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TACCTGAGGTCTACTTGTGTCTCCC[C/T]CCTTGTTCTCTGCCCCCAGACCATG
- Phenotype
-
MIM: 600950
MIM: 614404
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
AANAT
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs111924263] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- AANAT
- Gene Name
- aralkylamine N-acetyltransferase
- Gene
- RHBDF2
- Gene Name
- rhomboid 5 homolog 2
View Full Product Details