Product Details

SNP ID
rs2283143
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:238058 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ATGGTTTGTGTTGGATAGCACTTGG[T/C]AATTTTCAAAGCATTTTATCTGCGT
Phenotype
MIM: 603481 MIM: 604481
Polymorphism
T/C, Transition substitution
Allele Nomenclature
Literature Links
PSMD13 PubMed Links

Gene Details

Gene
PSMD13
Gene Name
proteasome 26S subunit, non-ATPase 13
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002817.3 Intron NP_002808.3
NM_175932.2 Intron NP_787128.2
XM_011520235.2 Intron XP_011518537.1
Gene
SIRT3
Gene Name
sirtuin 3
There are no transcripts associated with this gene.

View Full Product Details