Product Details

SNP ID
rs2272208
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.18:23452945 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
CTCCAGGAAATAATGAATGAACAGA[A/G]AGAGCCTAGTTTGTTTATTTATTTA
Phenotype
MIM: 603579
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
RIOK3 PubMed Links

Gene Details

Gene
RIOK3
Gene Name
RIO kinase 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003831.3 123 UTR 5 NP_003822.2
XM_011526242.1 123 UTR 5 XP_011524544.1
XM_011526243.1 123 UTR 5 XP_011524545.1

View Full Product Details