Product Details

SNP ID
rs2279130
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:75266141 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GTGCACGAGAGTGGAGCCGGAGCCA[C/T]GTGGAGGCAGGGCCGGCCCGTCTAT
Phenotype
MIM: 107940
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ARRB1 PubMed Links

Gene Details

Gene
ARRB1
Gene Name
arrestin beta 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004041.4 1415 UTR 3 NP_004032.2
NM_020251.3 1415 UTR 3 NP_064647.1
XM_011545034.2 1415 UTR 3 XP_011543336.1
XM_011545035.2 1415 UTR 3 XP_011543337.1
XM_017017750.1 1415 UTR 3 XP_016873239.1
XM_017017751.1 1415 UTR 3 XP_016873240.1
XM_017017752.1 1415 UTR 3 XP_016873241.1
XM_017017753.1 1415 UTR 3 XP_016873242.1
XM_017017754.1 1415 UTR 3 XP_016873243.1

View Full Product Details