Product Details

SNP ID
rs2516562
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:135947020 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAAGAGCCCGAAGCAACCAGTTCAC[A/G]TGAACATCAGAGTGACAATGAGTTA
Phenotype
MIM: 609087 MIM: 602882
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
FBXL21 PubMed Links

Gene Details

Gene
FBXL21
Gene Name
F-box and leucine rich repeat protein 21 (gene/pseudogene)
There are no transcripts associated with this gene.

Gene
LECT2
Gene Name
leukocyte cell derived chemotaxin 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002302.2 968 UTR 3 NP_002293.2

View Full Product Details