Product Details

SNP ID
rs2221236
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:159077416 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ACATGTGGTGAAGTTTTGGTTGCAG[C/T]ATTGTTTGTGTTATTAAAAATCCAT
Phenotype
MIM: 611622
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
IQCJ PubMed Links

Gene Details

Gene
IQCJ
Gene Name
IQ motif containing J
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001042705.2 Intron NP_001036170.1
NM_001042706.2 Intron NP_001036171.1
NM_001197100.1 Intron NP_001184029.1
Gene
IQCJ-SCHIP1
Gene Name
IQCJ-SCHIP1 readthrough
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001197113.1 Intron NP_001184042.1
NM_001197114.1 Intron NP_001184043.1

View Full Product Details