Product Details

SNP ID
rs3026076
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:66757157 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCTTGTGAAAGAGATGTAAGAATCA[A/C]TACTTTCTCAAGGTTCCTTTCTAAA
Phenotype
MIM: 611406
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
DYNC1LI2 PubMed Links

Gene Details

Gene
DYNC1LI2
Gene Name
dynein cytoplasmic 1 light intermediate chain 2
There are no transcripts associated with this gene.

Gene
LOC106699570
Gene Name
uncharacterized LOC106699570
There are no transcripts associated with this gene.

Gene
TERB1
Gene Name
telomere repeat binding bouquet formation protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001136505.1 2022 Intron NP_001129977.1
XM_011523004.2 2022 Intron XP_011521306.1
XM_011523005.2 2022 Intron XP_011521307.1
XM_011523006.2 2022 Intron XP_011521308.1
XM_011523007.2 2022 Intron XP_011521309.1
XM_011523008.2 2022 Intron XP_011521310.1
XM_011523009.2 2022 Intron XP_011521311.1
XM_011523010.2 2022 Intron XP_011521312.1
XM_011523011.2 2022 Intron XP_011521313.1
XM_011523012.2 2022 Intron XP_011521314.1
XM_011523014.2 2022 Intron XP_011521316.1
XM_011523015.2 2022 Intron XP_011521317.1
XM_011523016.2 2022 UTR 3 XP_011521318.1
XM_011523017.1 2022 Intron XP_011521319.1
XM_011523018.1 2022 Intron XP_011521320.1
XM_011523020.1 2022 Intron XP_011521322.1
XM_017023157.1 2022 Intron XP_016878646.1
XM_017023158.1 2022 UTR 3 XP_016878647.1
XM_017023159.1 2022 Intron XP_016878648.1

View Full Product Details