Product Details

SNP ID
rs370425026
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:54219113 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CACCCTCGGTCGACCCATGGGTCCC[C/G]CGCTTCCCTACTCACCAGATGTCCT
Phenotype
MIM: 604820
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
LILRB3 PubMed Links

Gene Details

Gene
LILRB3
Gene Name
leukocyte immunoglobulin like receptor B3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001081450.2 Intron NP_001074919.2
NM_001320960.1 Intron NP_001307889.1
NM_006864.3 Intron NP_006855.3
XM_011526381.2 Intron XP_011524683.1
XM_011526382.2 Intron XP_011524684.1
XM_017026218.1 Intron XP_016881707.1
XM_017026219.1 Intron XP_016881708.1
XM_017026220.1 Intron XP_016881709.1
XM_017026221.1 Intron XP_016881710.1
XM_017026222.1 Intron XP_016881711.1
XM_017026223.1 Intron XP_016881712.1

View Full Product Details