Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_004913.2 | Intron | NP_004904.2 | |||
XM_005256329.4 | Intron | XP_005256386.1 | |||
XM_005256330.4 | Intron | XP_005256387.1 | |||
XM_005256331.2 | Intron | XP_005256388.1 | |||
XM_006721350.2 | Intron | XP_006721413.1 | |||
XM_011523476.2 | Intron | XP_011521778.1 | |||
XM_011523477.2 | Intron | XP_011521779.1 | |||
XM_011523478.2 | Intron | XP_011521780.1 | |||
XM_011523479.2 | Intron | XP_011521781.1 | |||
XM_011523480.2 | Intron | XP_011521782.1 |