Product Details

SNP ID
rs35809332
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:117084302 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCAAAGATTATGAAGATTTCTCTTT[C/T]CTCTTTGGCGGACAGTCTACTCTAT
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
TRAM1L1 PubMed Links

Gene Details

Gene
TRAM1L1
Gene Name
translocation associated membrane protein 1-like 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_152402.2 1279 Silent Mutation AGA,AGG R364R NP_689615.2

View Full Product Details