Product Details

SNP ID
hCV25639520
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:104114119 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACCCTATCGTCGTCAGTCATGGCTA[C/G]CATCACTGCGTGTGTGGGTAACAGC
Phenotype
MIM: 300941
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
LOC286437 PubMed Links

Gene Details

Gene
LOC286437
Gene Name
uncharacterized LOC286437
There are no transcripts associated with this gene.

Gene
SLC25A53
Gene Name
solute carrier family 25 member 53
There are no transcripts associated with this gene.

Gene
ZCCHC18
Gene Name
zinc finger CCHC-type containing 18
There are no transcripts associated with this gene.

View Full Product Details