Product Details

SNP ID
rs116737608
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:108472909 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCTGTGAAATTAGTATACTTCTTCT[A/G]TTCTCTCTGTGCTTTTGGTCATACT
Phenotype
MIM: 612123
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
PNPLA8 PubMed Links

Gene Details

Gene
PNPLA8
Gene Name
patatin like phospholipase domain containing 8
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001256007.2 Intron NP_001242936.1
NM_001256008.2 Intron NP_001242937.1
NM_001256009.2 Intron NP_001242938.1
NM_001256010.2 Intron NP_001242939.1
NM_001256011.2 Intron NP_001242940.1
NM_015723.4 Intron NP_056538.1
XM_005250396.4 Intron XP_005250453.1
XM_011516274.2 Intron XP_011514576.1
XM_011516275.2 Intron XP_011514577.1
XM_017012261.1 Intron XP_016867750.1

View Full Product Details