Product Details

SNP ID
rs10000773
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:64281260 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CATATAAAGTCATAAGCAATCTCAT[A/T]AAATGGGATCATATTTATAAGAAAA
Phenotype
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
TECRL PubMed Links

Gene Details

Gene
TECRL
Gene Name
trans-2,3-enoyl-CoA reductase-like
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001010874.4 Intron NP_001010874.2
XM_005265662.4 Intron XP_005265719.1
XM_005265663.2 Intron XP_005265720.1
XM_005265664.2 Intron XP_005265721.1
XM_005265665.3 Intron XP_005265722.1
XM_017007959.1 Intron XP_016863448.1
XM_017007960.1 Intron XP_016863449.1

View Full Product Details