Product Details

SNP ID
rs112825147
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:10984387 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAGATGCACAAGGTAGGGATCCCTG[C/T]GCCCGCCTCGCACCTGCGGCCTCTG
Phenotype
MIM: 603254
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
SMARCA4 PubMed Links

Gene Details

Gene
SMARCA4
Gene Name
SWI/SNF related, matrix associated, actin dependent regulator of chromatin, subfamily a, member 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001128844.1 Intron NP_001122316.1
NM_001128845.1 Intron NP_001122317.1
NM_001128846.1 Intron NP_001122318.1
NM_001128847.1 Intron NP_001122319.1
NM_001128848.1 Intron NP_001122320.1
NM_001128849.1 Intron NP_001122321.1
NM_003072.3 Intron NP_003063.2
XM_006722845.2 Intron XP_006722908.1
XM_006722846.2 Intron XP_006722909.1
XM_011528198.1 Intron XP_011526500.1
XM_017027160.1 Intron XP_016882649.1
XM_017027161.1 Intron XP_016882650.1
XM_017027162.1 Intron XP_016882651.1
XM_017027163.1 Intron XP_016882652.1
XM_017027164.1 Intron XP_016882653.1
XM_017027165.1 Intron XP_016882654.1
XM_017027166.1 Intron XP_016882655.1
XM_017027167.1 Intron XP_016882656.1
XM_017027168.1 Intron XP_016882657.1

View Full Product Details